Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 delta Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB327100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327100 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327100 Supplier Novus Biologicals Supplier No. NBP299093PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD3 delta Polyclonal antibody specifically detects CD3 delta in Mouse samples. It is validated for Western Blot

Specifications

Antigen CD3 delta
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse CD3 delta.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 915
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.