Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 delta Rabbit anti-Human, Clone: 138, Novus Biologicals™
SDP

Catalog No. NB020942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020942 50 μg
NB020941 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020942 Supplier Novus Biologicals Supplier No. NBP28973050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD3 delta Monoclonal specifically detects CD3 delta in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CD3 delta
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 138
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Host Species Rabbit
Immunogen Recombinant Human CD3 delta protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 915
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.