Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 epsilon Antibody (145-2C11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB368925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368925 Supplier Novus Biologicals Supplier No. FAB484Z
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Cell depletion, CyTOF, Flow Cytometry, Immunoprecipitation, T Cell Stimulation.

Specifications

Antigen CD3 epsilon
Applications Cell Depletion, CyTOF, Flow Cytometry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 145-2C11
Conjugate DyLight 755
Dilution Flow Cytometry, Immunoprecipitation, Cell depletion, T Cell Stimulation, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Host Species Hamster
Immunogen H-2Kb-specific mouse cytotoxic T-lymphocyte
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.