| Antigen |
CD3 epsilon |
| Applications |
Western Blot, CyTOF, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification |
Polyclonal |
| Concentration |
1.0 mg/mL |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 1 - 2 ug/mL, CyTOF-ready |
| Formulation |
10 mM PBS with 0.05% Sodium Azide |
| Gene Alias |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE |
| Gene Symbols |
CD3E |
| Host Species |
Rabbit |
| Immunogen |
A synthetic peptide aa156-168 (ERPPPVPNPDYEP) from human CD3 epsilon (Uniprot: P07766) |
| Molecular Weight of Antigen |
23 kDa |
| Purification Method |
Protein A purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
916 |
| Test Specificity |
Recognizes the epsilon-chain of CD3, which consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta) with MW ranging from 16-28kDa. The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). Reportedly, CD3 complex is involved in signal transduction to the T cell interior following antigen recognition. The CD3 antigen is first detectable in early thymocytes and probably represents one of the earliest signs of commitment to the T cell lineage. In cortical thymocytes, CD3 is predominantly intra-cytoplasmic. However, in medullary thymocytes, it appears on the T cell surface. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms. |
| Target Species |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Equine, Feline, Guinea Pig, Hamster, Other, Primate |
| Content And Storage |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Form |
Purified |
| Isotype |
IgG |
|
Show More
Show Less
|
|