Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 epsilon Antibody (PPT3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128227 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128227 Supplier Novus Biologicals Supplier No. NBP128227
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Porcine samples. It is validated for Flow Cytometry.

Specifications

Antigen CD3 epsilon
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone PPT3
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 1 ug/10^6 cells
Gene Alias CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Gene Symbols CD3E
Host Species Mouse
Immunogen Purified CD3 molecules from porcine thymus
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 916
Test Specificity Porcine CD3 epsilon, Mr 20 kDa
Target Species Pig
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.