Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: SP34-2, BD Horizon™
Mouse Monoclonal Antibody
$190.50 - $458.00
Specifications
| Antigen | CD3 |
|---|---|
| Clone | SP34-2 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD3 Monoclonal antibody specifically detects CD3 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.Specifications
| CD3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 λ | |
| Refrigerate (2 to 8°C) |
| SP34-2 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Baboon, Cynomolgus, Rhesus Macaque, Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title