Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 zeta Antibody (H146-968), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP144683IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144683IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144683IR Supplier Novus Biologicals Supplier No. NBP144683IR
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD3 ζ
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H146-968
Conjugate DyLight 755
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 919
Test Specificity The hamster antibody H146-968 reacts with CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells.
Target Species Human, Mouse
Form Purified
Isotype IgG2 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.