Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 zeta Antibody (H146-968), FITC, Novus Biologicals™
SDP

Catalog No. NB358815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358815 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358815 Supplier Novus Biologicals Supplier No. NBP144683F
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF

Specifications

Antigen CD3 zeta
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone H146-968
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Host Species Armenian Hamster
Immunogen Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 919
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.