Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 zeta, p Tyr111 Antibody (EM-55), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP237709A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP237709A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP237709A32 Supplier Novus Biologicals Supplier No. NBP237709AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).

Specifications

Antigen CD3 zeta
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone EM-55
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Gene Symbols CD247
Host Species Mouse
Immunogen BP1 (pTyr111/123) - KLH.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 919
Test Specificity Antibody recognizes phosphorylated tyrosine 111 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.