Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Ligand/TNFSF8 Antibody (MM0168-3G56), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP211973G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211973G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211973G Supplier Novus Biologicals Supplier No. NBP211973G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD30 Ligand/TNFSF8 Monoclonal specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD30 Ligand/TNFSF8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0168-3G56
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD153, CD153 antigen, CD30 antigen ligand, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Gene Symbols TNFSF8
Host Species Mouse
Immunogen Human recombinant CD30 Ligand/TNFSF8 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 944
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.