Learn More
Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody.
$245.00 - $1124.00
Specifications
| Antigen | CD30 Ab-1 |
|---|---|
| Clone | Ber-H2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|
MS361S0
|
Epredia™
MS361S0 |
100 μL | Concentrated |
Each for $245.00
|
|
||||
|
MS361S
|
Epredia™
MS361S |
1 mL | Concentrated |
Each for $1,124.00
|
|
||||
|
MS361R7
|
Epredia™
MS361R7 |
7 mL | Prediluted |
Each for $545.00
|
|
||||
|
MS361RQ
|
Epredia™
MS361RQ |
12 mL | Ready-to-Use |
Each for $565.00
|
|
||||
|
MS361S1
|
Epredia™
MS361S1 |
500 μL | Concentrated | N/A | N/A | N/A | |||
|
MS361R1
|
Epredia™
MS361R1 |
1 mL | Prediluted | N/A | N/A | N/A | |||
Description
CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells.Host Species: Mouse
Clone: Ber-H2
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with baboon and monkey. Others not known.
Epitope: Not determined
Immunogen: Co cells
Molecular Weight: 120 kDa (mature), 95 kDa (precursor)
Positive Control: Hodgkin's lymphoma
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD30 Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Co cells |
| Ber-H2 | |
| Monoclonal | |
| Mouse | |
| Oncogenesis | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.