Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30/TNFRSF8 Antibody (Ki-1/1747R) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254556100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254556100 100 μg
NB254556200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254556100 Supplier Novus Biologicals Supplier No. NBP254556100UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen CD30/TNFRSF8
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone Ki-1/1747R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Gene Symbols TNFRSF8
Host Species Rabbit
Immunogen Recombinant human full-length TNFRSF8 protein
Molecular Weight of Antigen 113 kDa
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Embryonic Stem Cell Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 943
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.