Learn More
CD30/TNFRSF8 Antibody (Ki-3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD30/TNFRSF8 |
| Applications | CyTOF, ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | Ki-3 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Gene Alias | CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 |
| Gene Symbols | TNFRSF8 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.