Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30/TNFRSF8 Antibody (Ki-3) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$268.00 - $584.50

Specifications

Antigen CD30/TNFRSF8
Clone Ki-3
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25268225
SDP
View Documents
Novus Biologicals
NBP2526820.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP25268202
SDP
View Documents
Novus Biologicals
NBP2526820.2MG
0.2 mg
Each for $584.50
Only null left
Add to Cart
 
Description

Description

CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Functional, Immunofluorescence, CyTOF-ready.
Specifications

Specifications

CD30/TNFRSF8
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Monoclonal
Purified
RUO
Human
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
TNFRSF8
IgG
Protein A or G purified
The antibody binds to CD30.
Ki-3
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, CyTOF
Unconjugated
Rabbit
Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Embryonic Stem Cell Markers, Immunology, Stem Cell Markers
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
943
Human CD30.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.