Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30/TNFRSF8 Antibody (Ki-3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP52682549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP52682549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP52682549 Supplier Novus Biologicals Supplier No. NBP252682JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen CD30/TNFRSF8
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone Ki-3
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Gene Symbols TNFRSF8
Host Species Rabbit
Immunogen Human CD30.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Embryonic Stem Cell Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 943
Test Specificity The antibody binds to CD30.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.