Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300a/LMIR1 Antibody (172224) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB400722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400722 Supplier Novus Biologicals Supplier No. FAB1186E
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CD300a/LMIR1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 172224
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1, Leu28-Arg183, Accession # BAC80268
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11314
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.