Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300a/LMIR1 Antibody (MEM-260), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N1058726P59 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1058726P59 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1058726P59 Supplier Novus Biologicals Supplier No. NB11058726PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300a/LMIR1
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-260
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen
Gene Symbols CD300A
Host Species Mouse
Immunogen HPB acute lymphoid leukemia cell line (NM_007261.2).
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11314.0
Test Specificity This antibody (clone MEM-260) reacts with CD300a, a 60 kDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.