Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300b/LMIR5/CD300LB Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167177 100 μg
NB167178 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167177 Supplier Novus Biologicals Supplier No. NBP316041100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD300b/LMIR5/CD300LB Polyclonal antibody specifically detects CD300b/LMIR5/CD300LB in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen CD300b/LMIR5/CD300LB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, TrIgGering receptor expressed on myeloid cells 5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human CD300b/LMIR5/CD300LB (NP_777552.3). MWLPPALLLLSLSGCFSIQGPESVRAPEQGSLTVQCHYKQGWETYIKWWCRGVRWDTCKILIETRGSEQGEKSDRVSIKDNQKDRTFTVTMEGLRRDDADVYWCGIERRGPDLGTQVKVIVDPEGAASTTASSPTNSNMAVFIGSHKRNHYMLLVFVKVPILLILVTAILWLKGSQRVPEEPGEQPIYMNFSEPLTKDMAT
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124599
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.