Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300b/LMIR5/CD300LB Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $389.00
Specifications
Antigen | CD300b/LMIR5/CD300LB |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD300b/LMIR5/CD300LB Polyclonal antibody specifically detects CD300b/LMIR5/CD300LB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CD300b/LMIR5/CD300LB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, TrIgGering receptor expressed on myeloid cells 5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human CD300b/LMIR5/CD300LB (NP_777552.3). MWLPPALLLLSLSGCFSIQGPESVRAPEQGSLTVQCHYKQGWETYIKWWCRGVRWDTCKILIETRGSEQGEKSDRVSIKDNQKDRTFTVTMEGLRRDDADVYWCGIERRGPDLGTQVKVIVDPEGAASTTASSPTNSNMAVFIGSHKRNHYMLLVFVKVPILLILVTAILWLKGSQRVPEEPGEQPIYMNFSEPLTKDMAT | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, 50% glycerol, pH7.3 | |
124599 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title