Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300c Antibody (2799A), PerCP, Novus Biologicals™
SDP

Catalog No. NB304056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304056 Supplier Novus Biologicals Supplier No. FAB3256C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD300c Monoclonal specifically detects CD300c in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300c
Applications Flow Cytometry
Classification Monoclonal
Clone 2799A
Conjugate PerCP
Formulation PBS
Gene Alias CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR
Host Species Rabbit
Immunogen NS0-derived human CD300c protein, Met29-Arg183, Accession # Q08708
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10871
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.