Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300c Rabbit anti-Human, Clone: 72, Novus Biologicals™
SDP

Catalog No. NB021446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021446 50 μg
NB021445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021446 Supplier Novus Biologicals Supplier No. NBP28999150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD300c Monoclonal specifically detects CD300c in Human samples. It is validated for ELISA, Sandwich ELISA Capture.

Specifications

Antigen CD300c
Applications ELISA
Classification Monoclonal
Clone 72
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR
Host Species Rabbit
Immunogen Recombinant Human CD300c/LMIR2 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10871
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.