Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300e/LMIR6 Antibody (233810), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB313322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313322 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313322 Supplier Novus Biologicals Supplier No. FAB2705PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD300e/LMIR6 Monoclonal specifically detects CD300e/LMIR6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300e/LMIR6
Applications Flow Cytometry
Classification Monoclonal
Clone 233810
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CD300e/LMIR6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 342510
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.