Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300f/LMIR3/CD300LF Antibody (234903), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313259 Supplier Novus Biologicals Supplier No. FAB2774PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD300f/LMIR3/CD300LF Monoclonal specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300f/LMIR3/CD300LF
Applications Flow Cytometry
Classification Monoclonal
Clone 234903
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CLM1, CLM-1, IgSF13, IREM1, IREM-1, NKIR
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CD300f/LMIR3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.