Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300LG/Nepmucin Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB081296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081296 Supplier Novus Biologicals Supplier No. NBP298158V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD300LG/Nepmucin Polyclonal specifically detects CD300LG/Nepmucin in Rat samples. It is validated for ELISA.

Specifications

Antigen CD300LG/Nepmucin
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CD300LG/Nepmucin (XP_002724611.1; Met1-Arg160).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146894
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.