Learn More
CD301b (MGL2) Monoclonal Antibody (11A10-B7), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$346.50 - $1562.00
Specifications
Antigen | CD301b (MGL2) |
---|---|
Clone | 11A10-B7 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-301-182
![]() |
Invitrogen™
14301182 |
100 μg |
Each
for $346.50
Save 25% Reg : |
![]() |
|||||
14-301-137
![]() |
Invitrogen™
14301137 |
2 mg |
Each for $1,562.00
|
|
|||||
Description
Description: The monoclonal antibody 11A10-B7 recognizes mouse CD301b. CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming. Applications Reported: This 11A10-B7 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, and microscopy. Applications Tested: This 11A10-B7 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.Specifications
CD301b (MGL2) | |
0.5 mg/mL | |
Unconjugated | |
Rat | |
Mouse | |
CD301; CD301b; macrophage galactose N-acetyl-galactosamine specific lectin 2; macrophage galactose-type C-type lectin 2; Mgl2 | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
11A10-B7 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
216864 | |
Primary | |
4°C | |
Mgl2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.