Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD302/CLEC13A Antibody (071), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB316254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316254 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316254 Supplier Novus Biologicals Supplier No. NBP290687MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD302/CLEC13A Monoclonal antibody specifically detects CD302/CLEC13A in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen CD302/CLEC13A
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 071
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BIMLEC, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD302/CLEC13A (Uniprot#: Q9DCG2-2; Met1-His156).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9936
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.