Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 Antibody (377537) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB372380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372380 Supplier Novus Biologicals Supplier No. FAB33871A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CD31/PECAM-1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 377537
Conjugate APC
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1, Gln28-Lys602 (predicted), Accession # Q95242
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5175
Target Species Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.