Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 Antibody (C31.3 + C31.7 + C31.10) - IHC-Prediluted, Novus Biologicals™
SDP

Catalog No. NBP248331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248331 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP248331 Supplier Novus Biologicals Supplier No. NBP248331
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Rat, Cynomolgus Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD31/PECAM-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C31.3 + C31.7 + C31.10
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Gene Symbols PECAM1
Host Species Mouse
Immunogen Recombinant full-length human CD31/PECAM-1 protein (Uniprot: P16284)
Molecular Weight of Antigen 82.5 kDa
Purification Method Protein A or G purified
Quantity 7 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5175
Test Specificity CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 monoclonal antibody reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence.
Target Species Human, Rat, Cynomolgus Monkey, Rabbit
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.