Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (LCI-4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100653360.025MG
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q95242 | |
| PECAM1 | |
| Porcine CD31/human IgGFc fusion protein | |
| 0.025 mg | |
| Angiogenesis, Cancer, Cellular Markers, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 5175 | |
| Human, Pig | |
| Purified |
| Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| LCI-4 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes porcine CD31, also known as Platelet endothelial cell adhesion molecule (PECAM-1). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction