Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (LCI-4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $622.00
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | LCI-4 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017710
![]() |
Novus Biologicals
NB100653360.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NB100653361
![]() |
Novus Biologicals
NB100653360.1MG |
0.1 mg |
Each for $622.00
|
|
|||||
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| CD31/PECAM-1 | |
| 1.0 mg/mL | |
| Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cellular Markers, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| IgG1 | |
| Protein G purified | |
| Recognizes porcine CD31, also known as Platelet endothelial cell adhesion molecule (PECAM-1). |
| LCI-4 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Pig | |
| Q95242 | |
| 5175 | |
| Porcine CD31/human IgGFc fusion protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title