Learn More
Description
Specifications
Specifications
| Antigen | CD31/PECAM-1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM532 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Gene Alias | adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule |
| Gene Symbols | PECAM1 |
| Host Species | Mouse |
| Immunogen | Human recombinant CD31/PECAM-1 protein (Uniprot: P16284) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
