| Antigen |
CD31/PECAM-1 |
| Applications |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
C31.3 + JC/70A |
| Conjugate |
Unconjugated |
| Dilution |
Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml |
| Formulation |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. |
P16284 |
| Gene Alias |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule |
| Gene Symbols |
PECAM1 |
| Host Species |
Mouse |
| Immunogen |
Human recombinant CD31 protein (C31.3) & Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A) |
| Purification Method |
Protein A or G purified |
| Quantity |
0.02 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
5175 |
| Test Specificity |
CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence. |
| Target Species |
Human, Primate, Rabbit |
| Content And Storage |
Store at 4C. |
| Form |
Purified |
| Isotype |
IgG |
|
Show More
Show Less
|
|