Learn More
CD314 (NKG2D) Monoclonal Antibody (5C6), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$1562.00
Specifications
| Antigen | CD314 (NKG2D) |
|---|---|
| Clone | 5C6 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-587-937
|
Invitrogen™
14587937 |
2 mg |
Each of 1 for $1,562.00
|
|
|||||
Description
The 5C6 MAb reacts with the human NKG2D, a 42 kDa lectin-like molecule expressed by NK cells, gammadelta T cells, some CD4^+ and CD8^+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gammadelta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.Specifications
| CD314 (NKG2D) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D | |
| IgG2a | |
| Affinity chromatography | |
| Antibody |
| 5C6 | |
| Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P26718 | |
| 22914 | |
| Primary | |
| 4°C | |
| Klrk1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.