Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD325 (N-Cadherin) Monoclonal Antibody (8C11), PE-Cyanine7, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 25325942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-325-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-325-942 Supplier Invitrogen™ Supplier No. 25325942
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 8C11 monoclonal antibody reacts with human CD325, also known as N-Cadherin. CD325 is a 130 kDa member of the Cadherin superfamily, and consists of five extracellular repeats, a transmembrane domain and a cytoplasmic domain. CD325 deficient mice die at

N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD325 (N-Cadherin)
Applications Flow Cytometry
Classification Monoclonal
Clone 8C11
Concentration 5 μL/Test
Conjugate PE-Cyanine7
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CDH2
Gene Accession No. P19022
Gene Alias bib; cadherin 2; cadherin 2 L homeolog; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, neuronal; cadherin 2, type 1 preproprotein; cadherin 2, type 1, N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal) L homeolog; cadherin 2, type 1, N-cadherin a (neuronal); Cadherin2; cadherin-2; Cadherin-2A; Cadherin-N; calcium-dependent adhesion protein, neuronal; CD325; CDH2; cdh2.L; cdh2a; cdh2-a; cdh2-b; CDHN; cdhn-a; CDw325; CDw325 antigen; cell adhesion molecule; glass onion; glo; I79_014914; labyrinth; lyr; NCAD; N-Cad; N-cadherin; N-cadherin 1; N-cadherin A; N-cadherin precursor; N-cadherin-2; neural cadherin; Neural cadherin A; neural cadherin-2; neural calcium-dependent cell adhesion molecule; neural-cadherin; pac; Parachute; unnamed protein product; wu:fb47h04; XELAEV_18031953mg; ZNCAD
Gene Symbols CDH2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.