Learn More
CD325 (N-Cadherin) Monoclonal Antibody (8C11), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$463.00
Specifications
| Antigen | CD325 (N-Cadherin) |
|---|---|
| Clone | 8C11 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25-325-942
![]()
|
Invitrogen™
25325942 |
100 Tests |
Each of 1 for $463.00
|
|
|||||
Description
The 8C11 monoclonal antibody reacts with human CD325, also known as N-Cadherin. CD325 is a 130 kDa member of the Cadherin superfamily, and consists of five extracellular repeats, a transmembrane domain and a cytoplasmic domain. CD325 deficient mice die at
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.Specifications
| CD325 (N-Cadherin) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P19022 | |
| CDH2 | |
| Primary | |
| Antibody |
| 8C11 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| Mouse | |
| Human | |
| 1000 | |
| IgG1 κ | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CDH2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.