Learn More
CD33 Mouse anti-Human, FITC, Clone: HIM3-4, BD
Mouse Monoclonal Antibody
$104.50 - $231.00
Specifications
Antigen | CD33 |
---|---|
Clone | HIM3-4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Description
Reacts with a 67 kDa type I transmembrane glycoprotein expressed on monocytes, activated T cells, myeloid progenitors as well as mast cells. CD33 is absent on normal platelets, lymphocytes, erythrocytes and hematopoietic stem cells. Reports indicate that this glycoprotein can function as a sialic acid-dependent cell adhesion molecule. This function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane. HIM3-4 reacts with human and monkey granulocytes and monocytes.
Specifications
CD33 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C |
HIM3-4 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.