Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD336 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CD336 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5996
![]() ![]() |
Proteintech Group Inc
551181AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to CD336.
CD336 is also known as NKp44 or NCR2. CD336 is a 44-kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.Specifications
CD336 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O95944 | |
9436 | |
Peptide | |
Primary | |
-20°C | |
NCR2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD336, LY95, Lymphocyte antigen 95 homolog, NCR2, NK cell activating receptor, NK p44, NKP44 | |
NCR2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title