Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Antibody (B-G25), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB388411 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388411 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388411 Supplier Novus Biologicals Supplier No. NBP314591IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone B-G25
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Host Species Mouse
Immunogen KG-1a cell line
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 947
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.