Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD34 Antibody (HPCA1/1806R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP253353100UG
Description
CD34 Monoclonal specifically detects CD34 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD34 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
Rabbit | |
95 kDa | |
100 μg | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
947 | |
Store at 4C. | |
IgG κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HPCA1/1806R | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
CD34 | |
Recombinant full-length human HPCA1 protein | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction