Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Antibody (HPCA1/1806R), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$316.50 - $574.50

Specifications

Antigen CD34
Clone HPCA1/1806R
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25335320
SDP
View Documents
Novus Biologicals
NBP25335320UG
20 μg
Each for $316.50
Only null left
Add to Cart
 
NB253353100
SDP
View Documents
Novus Biologicals
NBP253353100UG
100 μg
Each for $574.50
Only null left
Add to Cart
 
NB253353200
SDP
View Documents
Novus Biologicals
NBP253353200UG
200 μg N/A N/A N/A
Description

Description

CD34 Monoclonal specifically detects CD34 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD34
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
947
Recombinant full-length human HPCA1 protein
Primary
Store at 4C.
HPCA1/1806R
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human, Rat
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
CD34
IgG κ
Protein A or G purified
95 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.