Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Antibody (OTI11E1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270375R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270375R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270375R Supplier Novus Biologicals Supplier No. NBP270375R

Mouse Monoclonal Antibody

CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CD34
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI11E1
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Gene Symbols CD34
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 312-385 of human CD34 (NP_001020280) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 947.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.