Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Antibody (7G6) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25266702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25266702 0.2 mg
NBP25266725 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25266702 Supplier Novus Biologicals Supplier No. NBP2526670.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD35 Monoclonal specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready, Block/Neutralize.

Specifications

Antigen CD35
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Blocking Assay, CyTOF
Classification Monoclonal
Clone 7G6
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Gene Symbols CR1
Host Species Rabbit
Immunogen Purified Mouse CR1.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Test Specificity Recognises an epitope shared by 145-150kDa and 190kDa complement receptor proteins, originally designated CR2 (CD21) and CR1 (CD35).
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.