Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Antibody (7G6), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB343219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343219 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343219 Supplier Novus Biologicals Supplier No. NBP252667DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, CyTOF, Neutralization

Specifications

Antigen CD35
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7G6
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Host Species Rabbit
Immunogen Purified Mouse CR1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1378
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.