Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Antibody (E11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP233080W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233080W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233080W Supplier Novus Biologicals Supplier No. NBP233080AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD35 Monoclonal specifically detects CD35 in Human, Monkey, Baboon, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CD35
Applications ELISA
Classification Monoclonal
Clone E11
Conjugate Alexa Fluor 405
Dilution ELISA
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Gene Symbols CR1
Host Species Mouse
Immunogen Intact human monocytes were used as immunogen to generate the CD35 antibody (Hogg et al, 1984).
Molecular Weight of Antigen 215 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Test Specificity Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This monoclonal antibody is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This monoclonal antibody is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. They are CD21+/CD35+/CD1a-. This monoclonal antibody labels follicular dendritic cells and follicular dendritic cell sarcoma.
Target Species Human, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.