Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (E11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$524.00
Specifications
Antigen | CD35 |
---|---|
Clone | E11 |
Concentration | 0.2 mg/ml |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Immunoprecipitation |
Classification | Monoclonal |
Description
CD35 Monoclonal specifically detects CD35 in Human, Monkey, Baboon, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD35 | |
0.2 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
P17927 | |
1378 | |
Intact human monocytes were used as immunogen to generate the CD35 antibody (Hogg et al, 1984). | |
Primary | |
Store at 4C. | |
215 kDa |
E11 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Monkey | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
CR1 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This monoclonal antibody is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This monoclonal antibody is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. They are CD21+/CD35+/CD1a-. This monoclonal antibody labels follicular dendritic cells and follicular dendritic cell sarcoma. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title