Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Antibody (To5), HRP, Novus Biologicals™
SDP

Catalog No. NBP250177H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250177H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250177H Supplier Novus Biologicals Supplier No. NBP250177H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD35 Monoclonal specifically detects CD35 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CD35
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone To5
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Gene Symbols CR1
Host Species Mouse
Immunogen C3b receptor-enriched precipitates from tonsil cell membranes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.