Learn More
CD357 (AITR/GITR) Monoclonal Antibody (DTA-1), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$3260.00
Specifications
Antigen | CD357 (AITR/GITR) |
---|---|
Clone | DTA-1 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-587-438
![]() |
Invitrogen™
16587438 |
5 mg |
Each of 1 for $3,260.00
|
|
|||||
Description
The DTA-1 MAb reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naive mice, GITR is expressed predominantly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.Specifications
CD357 (AITR/GITR) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
21936 | |
Primary | |
4°C | |
Tnfrsf18 |
DTA-1 | |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
Functional Grade | |
Rat | |
Mouse | |
O35714 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.