| Antigen |
CD36/SR-B3 |
| Applications |
Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification |
Monoclonal |
| Clone |
185-1G2 |
| Concentration |
1 mg/ml |
| Conjugate |
Unconjugated |
| Dilution |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Formulation |
10 mM PBS |
| Gene Alias |
CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor |
| Host Species |
Mouse |
| Immunogen |
Stimulated human leukocytes |
| Purification Method |
Protein A or G purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
948 |
| Test Specificity |
Recognizes a protein of 80kDa-90kDa, identified as CD36 (Workshop IV; Code P-26). Its epitope maps between aa155-183. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischemia observed in sickle cell disease and cerebral malaria. Note that 1-4% of Japanese and East Asia population lack CD36. This monoclonal antibody blocks adhesion of P. falciparum parasitized red blood cells to CD36 and strongly inhibits collagen-induced platelet aggregation. |
| Target Species |
Human |
| Content And Storage |
Store at 4°C. |
| Form |
Purified |
| Isotype |
IgG2a κ |
|
Show More
Show Less
|
|