Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (877302) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB19553
Description
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Negative), Immunocytochemistry.Specifications
| CD36/SR-B3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Flow Cytometry (Negative), Immunocytochemistry | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 877302 | |
| Unconjugated | |
| PBS with No Preservative | |
| CD36 | |
| Partial recombinant human CD36 protein (amino acids 30-439) [UniProt P16671] | |
| 100 μg | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction