Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody (877302) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$192.47 - $517.59
Specifications
| Antigen | CD36/SR-B3 |
|---|---|
| Clone | 877302 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Immunohistochemistry, Flow Cytometry (Negative), Immunocytochemistry |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB19553SP
![]() |
Novus Biologicals
MAB19553SP |
25 μg |
Each for $192.47
|
|
|||||
MAB19553
![]() |
Novus Biologicals
MAB19553 |
100 μg |
Each for $517.59
|
|
|||||
Description
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Negative), Immunocytochemistry.Specifications
| CD36/SR-B3 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| PBS with No Preservative | |
| 948 | |
| Partial recombinant human CD36 protein (amino acids 30-439) [UniProt P16671] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 877302 | |
| Western Blot, Immunohistochemistry, Flow Cytometry (Negative), Immunocytochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| CD36 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title